Anti-FBXO21 Rabbit Polyclonal Antibody (Biotin)

Leverantör: US Biological
USBI035554-BIOTINEA 9900 SEK
USBI035554-BIOTIN
Anti-FBXO21 Rabbit Polyclonal Antibody (Biotin)
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: FBXO21 (F-box protein 21)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR