|
|
ORDER
SPECIFIKATIONER
Antigen symbol | HLA-C |
Antigen name | major histocompatibility complex, class I, C |
Reaktivitet | Human |
Conjugation | AP (Alkaline Phosphatase) |
Clonality | Polyclonal |
Antikroppstyp | Primary |
Värd | Rabbit |
Isotype | IgG |
Western blot | Yes |
ELISA | Yes |
Aminosyretal | 65 to 93 |
Epitop | internal |
Rening | purified by Protein A chromatography |
Form | liquid |
Storage buffer | PBS, pH 7,2, contains ~0,09% sodium azide |
Förvaringstemperatur | Cool |
Shipping temperature | Ice |