Anti-HLA-DMB Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Leverantör: US Biological
USBI036618-APCEA 9900 SEK
USBI036618-APC
Anti-HLA-DMB Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: HLA-DMB (major histocompatibility complex, class II, DM beta)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human