Anti-HLA-DMB Rabbit Polyclonal Antibody (Biotin)
Leverantör: US Biological
USBI036618-BIOTINEA 9900 SEK
USBI036618-BIOTIN
Anti-HLA-DMB Rabbit Polyclonal Antibody (Biotin)
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: HLA-DMB (major histocompatibility complex, class II, DM beta)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human