Anti-HLA-DMB Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Leverantör: US Biological
USBI036618-PEEA 9900 SEK
USBI036618-PE
Anti-HLA-DMB Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: HLA-DMB (major histocompatibility complex, class II, DM beta)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human