Anti-HLA-DRB4 Rabbit Polyclonal Antibody (Biotin)
Leverantör: US Biological
USBI036625-BIOTINEA 9900 SEK
USBI036625-BIOTIN
Anti-HLA-DRB4 Rabbit Polyclonal Antibody (Biotin)
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: HLA-DRB4 (major histocompatibility complex, class II, DR beta 4)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human