Anti-HLA-DRB5 Rabbit Polyclonal Antibody

Leverantör: US Biological
USBI036626EA 6825 SEK
USBI036626
Anti-HLA-DRB5 Rabbit Polyclonal Antibody
Antikroppar
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: HLA-DRB5 (major histocompatibility complex, class II, DR beta 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR