Anti-HMBOX1 Rabbit Polyclonal Antibody (Biotin)
Leverantör: US Biological
USBI036630-BIOTINEA 9900 SEK
USBI036630-BIOTIN
Anti-HMBOX1 Rabbit Polyclonal Antibody (Biotin)
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: HMBOX1 (homeobox containing 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse