Anti-HMGB1P10 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Leverantör: US Biological
USBI036633-APEA 9900 SEK
USBI036633-AP
Anti-HMGB1P10 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikroppar
Do not freeze!
Type: Primary
Antigen: HMGB1P10 (high mobility group box 1 pseudogene 10)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human