Anti-HMGB1P10 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Leverantör: US Biological
USBI036633-APCEA 9900 SEK
USBI036633-APC
Anti-HMGB1P10 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: HMGB1P10 (high mobility group box 1 pseudogene 10)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human