Anti-HMGB1P10 Rabbit Polyclonal Antibody (Biotin)
Leverantör: US Biological
USBI036633-BIOTINEA 9900 SEK
USBI036633-BIOTIN
Anti-HMGB1P10 Rabbit Polyclonal Antibody (Biotin)
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Type: Primary
Antigen: HMGB1P10 (high mobility group box 1 pseudogene 10)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human