Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-HMGA2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverantör: US Biological
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: HMGA2 (high mobility group AT-hook 2)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse



Logga in för att se avtalspriser och tillgänglighet.
  Kom ihåg mig i den här enheten
 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
200 µl 200 µl USBI036635-AP
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol HMGA2
Antigen name High mobility group AT-hook 2
Reaktivitet Mouse, Human
Conjugation AP (Alkaline Phosphatase)
Clonality Polyclonal
Antikroppstyp Primary
Värd Rabbit
Isotype IgG
Western Blot Yes
ELISA Yes
Flödescytometri Yes
Aminosyretal 71 to 100
Epitop C-terminal
Rening purified by Protein A chromatography
Form liquid
Storage Buffer PBS, pH 7,2, contains ~0,09% sodium azide
Storage temperature Cool
Shipping Temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Benämning Artikel nummer Availability Enhet Ditt pris Price Per Antal