Anti-KCNK1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Leverantör: US Biological
USBI037333-APCEA 9900 SEK
USBI037333-APC
Anti-KCNK1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: KCNK1 (potassium channel, subfamily K, member 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR