Anti-MTHFSD Rabbit Polyclonal Antibody (Biotin)
Leverantör: US Biological
USBI038658-BIOTINEA 9900 SEK
USBI038658-BIOTIN
Anti-MTHFSD Rabbit Polyclonal Antibody (Biotin)
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: MTHFSD (methenyltetrahydrofolate synthetase domain containing)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human