Anti-MTM1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Leverantör: US Biological
USBI038662-APCEA 9900 SEK
USBI038662-APC
Anti-MTM1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: MTM1 (myotubularin 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse