Anti-MTM1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverantör: US Biological
USBI038662-HRPEA 9900 SEK
USBI038662-HRP
Anti-MTM1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: MTM1 (myotubularin 1)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse