Anti-MTMR14 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Leverantör: US Biological
USBI038665-PEEA 9900 SEK
USBI038665-PE
Anti-MTMR14 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: MTMR14 (myotubularin related protein 14)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human