Anti-SH2D5 Rabbit Polyclonal Antibody
Leverantör: United States Biological
USBI041685EA 6825 SEK
USBI041685
Anti-SH2D5 Rabbit Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: SH2D5 (SH2 domain containing 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse