Anti-EMD Rabbit polyclonal antibody
Leverantör: United States Biological
USBIE2257-01CEA 6825 SEK
USBIE2257-01C
Anti-EMD Rabbit polyclonal antibody
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100
Type: Primary
Antigen: EMD (emerin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human