Anti-EMR1 Rat monoclonal antibody [clone: 9E38]
Leverantör: United States Biological
USBIE2261-41EEA 9025 SEK
USBIE2261-41E
Anti-EMR1 Rat monoclonal antibody [clone: 9E38]
Antikroppar
Type: Primary
Antigen: EMR1 (egf-like module containing, mucin-like, hormone receptor-like 1)
Clonality: monoclonal
Clone: 9E+38
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse