Anti-EMR1 Rat monoclonal antibody Biotin [clone: 9E38]
USBIE2261-41E10EA 8325 SEK
USBIE2261-41E10
Anti-EMR1 Rat monoclonal antibody Biotin [clone: 9E38]
Recommended dilution: Flow Cytometry: 1:10-1:50;
Type: Primary
Antigen: EMR1 (egf-like module containing, mucin-like, hormone receptor-like 1)
Clonality: monoclonal
Clone: 9E+38
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse