Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-EMR1 Rat monoclonal antibody unconjugated [clone: 9E38]

Leverantör: US Biological
Type: Primary
Antigen: EMR1 (egf-like module containing, mucin-like, hormone receptor-like 1)
Clonality: monoclonal
Clone: 9E+38
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse




 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
500 µg 500 µG USBIE2261-41E2
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol EMR1
Antigen name egf-like module containing, mucin-like, hormone receptor-like 1
Reaktivitet Mouse
Conjugation Unconjugated
Clonality Monoclonal
Antikroppstyp Primary
Clone 9E38
Värd Rat
Isotype IgG2b
Western blot Yes
ImmunoPrecipitation Yes
Flow cytometry Yes
Koncentration ~1 mg/ml
Rening purified by Protein G chromatography
Form liquid
Storage buffer PBS, pH 7,4
Förvaringstemperatur Cool
Shipping temperature Ice
Tested applications Radio Immunoassay
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping