Anti-EMR1 Rat monoclonal antibody [clone: BM8]
Leverantör: United States Biological
USBIE2261-41FEA 9575 SEK
USBIE2261-41F
Anti-EMR1 Rat monoclonal antibody [clone: BM8]
Antikroppar
Recommended dilution: Immunochemistry: 1:50.
Type: Primary
Antigen: EMR1 (egf-like module containing, mucin-like, hormone receptor-like 1)
Clonality: monoclonal
Clone: BM8
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse