Anti-EMX1 Rabbit Polyclonal Antibody (Biotin)
Leverantör: United States Biological
USBIE2265-05-BIOTEA 9900 SEK
USBIE2265-05-BIOT
Anti-EMX1 Rabbit Polyclonal Antibody (Biotin)
Antikroppar
Type: Primary
Antigen: EMX1 (empty spiracles homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human