Anti-EMX1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverantör: United States Biological
USBIE2265-05-HRPEA 9900 SEK
USBIE2265-05-HRP
Anti-EMX1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Type: Primary
Antigen: EMX1 (empty spiracles homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human