Anti-EN1 Rabbit polyclonal antibody
USBIE2270-01AEA 4400 SEK
USBIE2270-01A
Anti-EN1 Rabbit polyclonal antibody
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100
Type: Primary
Antigen: EN1 (engrailed homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse