Anti-EN1 Rabbit Polyclonal Antibody (Biotin)
Leverantör: United States Biological
USBIE2270-01-BIOTEA 7175 SEK
USBIE2270-01-BIOT
Anti-EN1 Rabbit Polyclonal Antibody (Biotin)
Antikroppar
Type: Primary
Antigen: EN1 (engrailed homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human