Anti-EN1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Leverantör: United States Biological
USBIE2270-01-PEEA 7175 SEK
USBIE2270-01-PE
Anti-EN1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50
Do not freeze!
Type: Primary
Antigen: EN1 (engrailed homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human