Vill du logga in igen?
Anti-EN2, CT-2 Rabbit polyclonal antibody Biotin
Leverantör: US Biological
Type: Primary
Antigen: EN2, CT-2
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Logga in för att se avtalspriser och tillgänglighet.
  Kom ihåg mig i den här enheten
ORDER
Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
200 µl 200 µl USBIE2270-10C-BIOT
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFIKATIONER
Antigen symbol EN2, CT-2
Reaktivitet Human
Conjugation Biotin
Clonality Polyclonal
Antikroppstyp Primary
Värd Rabbit
Isotype IgG
Western Blot Yes
ELISA Yes
Storage temperature Cool
Shipping Temperature Ice
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Benämning Artikel nummer Availability Enhet Ditt pris Price Per Antal