Anti-EN2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Leverantör: United States Biological
USBIE2270-10E-APCEA 9900 SEK
USBIE2270-10E-APC
Anti-EN2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: EN2 (engrailed homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse