Anti-GHRL Rabbit Polyclonal Antibody

Leverantör: United States Biological
USBIO4802-20DEA 7850 SEK
USBIO4802-20D
Anti-GHRL Rabbit Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 1:100000; Western Blot: 1-10 ?g/ml

Type: Primary
Antigen: GHRL (ghrelin/obestatin prepropeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR