Vill du logga in igen?
Anti-OCLN Guinea pig polyclonal antibody unconjugated
Leverantör: US Biological
Recommended dilution: Immunofluorescence: 1:50; Western Blot: 1:50; Immunochemistry: 1:50
Type: Primary
Antigen: OCLN (occludin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Guinea pig
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat
ORDER
Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
100 µg 100 µG USBIO5202-01
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFIKATIONER
Antigen symbol OCLN
Antigen name occludin
Reaktivitet Rat, Dog, Mouse, Human
Conjugation Unconjugated
Clonality Polyclonal
Antikroppstyp Primary
Värd Guinea Pig
Isotype IgG
Western blot Yes
Immunofluorescence Yes
Aminosyretal 358 to 504
Koncentration ~0,1 mg/ml
Rening purified by Protein A chromatography
Form liquid
Storage buffer PBS, 0,1% BSA, 0,02% sodium azide.
Förvaringstemperatur Cool
Shipping temperature Ice
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal