Vill du logga in igen?
Anti-SLC22A2 Rabbit polyclonal antibody unconjugated
Leverantör: US Biological
Recommended dilution: Immunochemistry: 1:25
Type: Primary
Antigen: SLC22A2 (solute carrier family 22 (organic cation transporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
ORDER
Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
250 µl 250 µl USBIO5401-08
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFIKATIONER
Antigen symbol Solute carrier family 22, member 2
Antigen name solute carrier family 22 (organic cation transporter), member 2
Reaktivitet Mouse, Human, Rat
Conjugation Unconjugated
Clonality Polyclonal
Antikroppstyp Primary
Värd Rabbit
Koncentration ~0,1 mg/ml
Rening purified by immunoaffinity chromatography
Form liquid
Storage buffer PBS, pH 7.6, 0,2% BSA and 15mM sodium azide, before the addition of glycerol to 40%
Förvaringstemperatur Cool
Shipping temperature Ice
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal