Vill du logga in igen?
Anti-SLC22A2 Mouse monoclonal antibody unconjugated [clone: MRQ-2]
Leverantör: US Biological
Recommended dilution: Immunochemistry: 1:25-1:100
Type: Primary
Antigen: SLC22A2 (solute carrier family 22 (organic cation transporter), member 2)
Clonality: monoclonal
Clone: MRQ-2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
ORDER
Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
1 ml 1 ml USBIO5401-08D
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFIKATIONER
Antigen symbol Solute carrier family 22, member 2
Antigen name solute carrier family 22 (organic cation transporter), member 2
Reaktivitet Human
Conjugation Unconjugated
Clonality Monoclonal
Antikroppstyp Primary
Clone MRQ-2
Värd Mouse
Isotype IgG1 kappa
Mått ascites
Form liquid
Storage buffer PBS, pH 7,4, protebase, sodium azide.
Förvaringstemperatur Cool
Shipping temperature Ice
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal