Anti-PAG3 Rabbit Polyclonal Antibody

Leverantör: United States Biological
USBIP1951-30EA 8075 SEK
USBIP1951-30
Anti-PAG3 Rabbit Polyclonal Antibody
Antikroppar
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: PAG3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR