Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-PTPRT Goat polyclonal antibody unconjugated

Leverantör: US Biological
Recommended dilution: ELISA: 1:32000; Western Blot: 0. 3-1 μg/ml

Type: Primary
Antigen: PTPRT (protein tyrosine phosphatase, receptor type, T)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse



Logga in för att se avtalspriser och tillgänglighet.
  Kom ihåg mig i den här enheten
 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
100 µg 100 µG USBIP9109-40T2
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol PTPRT
Antigen name Protein tyrosine phosphatase receptor type T
Reaktivitet Human, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antikroppstyp Primary
Värd Goat
Western Blot Yes
ELISA Yes
Koncentration ~0,5 mg/ml
Rening purified by immunoaffinity chromatography
Form liquid
Storage Buffer Tris saline, 0,02% sodium azide, pH 7,3, 0,5% BSA.
Storage temperature Cool
Shipping Temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Benämning Artikel nummer Availability Enhet Ditt pris Price Per Antal