Anti-PTPRT Goat Polyclonal Antibody

Leverantör: United States Biological
USBIP9109-40YEA 7350 SEK
USBIP9109-40Y
Anti-PTPRT Goat Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 1:32000; Western Blot: 0. 3-1 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: PTPRT (protein tyrosine phosphatase, receptor type, T)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR