Anti-PTP4A1 Goat Polyclonal Antibody

Leverantör: United States Biological
USBIP9109-45XEA 6400 SEK
USBIP9109-45X
Anti-PTP4A1 Goat Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 1:2000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: PTP4A1 (protein tyrosine phosphatase type IVA, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR