Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-PLRG1 Rabbit polyclonal antibody unconjugated

Leverantör: US Biological
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: PLRG1 (pleiotropic regulator 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse




 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
200 µl 200 µl USBIP9109-45Y
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol PLRG1
Antigen name Pleiotropic Regulator 1
Reaktivitet Mouse, Human
Conjugation Unconjugated
Clonality Polyclonal
Antikroppstyp Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Rening purified by Protein G chromatography
Form liquid
Storage buffer PBS, 0,09% sodium azide.
Förvaringstemperatur Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Benämning Artikel nummer Availability Enhet Ditt pris Price Per Antal