Anti-Protein Tyrosine Phosphatase Type 4A, Member 3, Isoform 2 Chicken Polyclonal Antibody
Leverantör: United States Biological
USBIP9109-48AEA 5525 SEK
USBIP9109-48A
Anti-Protein Tyrosine Phosphatase Type 4A, Member 3, Isoform 2 Chicken Polyclonal Antibody
Antikroppar
Recommended dilution: Western Blot: 1:1000-1:2000
Type: Primary
Antigen: Protein Tyrosine Phosphatase Type 4A, Member 3, Isoform 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human