Anti-PRL3 Rabbit Polyclonal Antibody
Leverantör: United States Biological
USBIP9109-48C2EA 6825 SEK
USBIP9109-48C2
Anti-PRL3 Rabbit Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Type: Primary
Antigen: PRL3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human