Anti-PRL3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Leverantör: United States Biological
USBIP9109-48C2-APCEA 9900 SEK
USBIP9109-48C2-APC
Anti-PRL3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: PRL3
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human