Anti-PRL3 Rabbit Polyclonal Antibody (Biotin)

Leverantör: United States Biological
USBIP9109-48C2BIOTEA 7475 SEK
USBIP9109-48C2BIOT
Anti-PRL3 Rabbit Polyclonal Antibody (Biotin)
Antikroppar
Type: Primary
Antigen: PRL3
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR