Anti-Prothrombin Burro Polyclonal Antibody
Leverantör: United States Biological
USBIP9115-05AEA 4000 SEK
USBIP9115-05A
Anti-Prothrombin Burro Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 10 ?g/ml; Western Blot: 10 ?g/ml
Type: Primary
Antigen: Prothrombin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Human