Vill du logga in igen?
Anti-Prothrombin Sheep polyclonal antibody unconjugated
Leverantör: US Biological
Type: Primary
Antigen: Prothrombin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
ORDER
Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
1 mg 1 mg USBIP9115-08F
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFIKATIONER
Antigen symbol Prothrombin
Reaktivitet Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antikroppstyp Primary
Värd Sheep
Isotype IgG
ELISA Yes
Koncentration ~10 mg/ml
Rening purified by Protein G chromatography
Form liquid
Storage buffer 0,05M sodium phosphate, 0,1M sodium chloride, 1mM EDTA, pH 6.6.
Förvaringstemperatur Cool
Shipping temperature Ice
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal