Vill du logga in igen?
Anti-PCDHA1 Mouse monoclonal antibody unconjugated [clone: 9G423]
Leverantör: US Biological
Type: Primary
Antigen: PCDHA1 (protocadherin alpha 1)
Clonality: monoclonal
Clone: 9G423
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
ORDER
Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
100 µg 100 µG USBIP9119-10E
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFIKATIONER
Antigen symbol PCDHA1
Antigen name Protocadherin alpha 1
Reaktivitet Human
Conjugation Unconjugated
Clonality Monoclonal
Antikroppstyp Primary
Clone 9G423
Värd Mouse
Isotype IgG
Western blot Yes
ELISA Yes
Rening purified by affinity chromatography
Form lyophilized powder
Storage buffer PBS, pH 7,2
Förvaringstemperatur Cool
Shipping temperature Ice
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal