Anti-PCDHA3 Mouse Polyclonal Antibody

Leverantör: United States Biological
USBIP9119-12EA 6450 SEK
USBIP9119-12
Anti-PCDHA3 Mouse Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: PCDHA3 (protocadherin alpha 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR