Anti-PCDHA5 Mouse Monoclonal Antibody [clone: 9G427]
Leverantör: United States Biological
USBIP9119-14EA 6300 SEK
USBIP9119-14
Anti-PCDHA5 Mouse Monoclonal Antibody [clone: 9G427]
Antikroppar
Type: Primary
Antigen: PCDHA5 (protocadherin alpha 5)
Clonality: monoclonal
Clone: 9G427
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human