Anti-SMAD6 Rabbit Polyclonal Antibody
Leverantör: United States Biological
USBIS1014-55H2EA 6275 SEK
USBIS1014-55H2
Anti-SMAD6 Rabbit Polyclonal Antibody
Antikroppar
Recommended dilution: Western Blot: 1:1000-1:2000
Type: Primary
Antigen: SMAD6 (SMAD family member 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Rat