Anti-SMAD6 Mouse Monoclonal Antibody [clone: 4F3]

Leverantör: United States Biological
USBIS1014-55H5EA 8175 SEK
USBIS1014-55H5
Anti-SMAD6 Mouse Monoclonal Antibody [clone: 4F3]
Antikroppar
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: SMAD6 (SMAD family member 6)
Clonality: Monoclonal
Clone: 4F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR